Human C5a, Tag Free (C5A-H5116) is expressed from E.coli cells. It contains AA Leu 679 - Arg 751 (Accession P01031). 1 * 1 mg
Katalog # ACRBC5A-H5116-1MG
Human C5a, Tag Free (C5A-H5116) is expressed from E.coli cells. It contains AA Leu 679 - Arg 751 (Accession P01031). 1 * 1 mg
Katalog # ACRBC5A-H5116-1MG