ContactUsLinkComponent
Recombinat Protein,Biotinylated Human CEACAM-1 CD66a Protein, Avitag™,His Tag,Biotinylated Human CEACAM-1, Avitag,His Tag (CE1-H82E 5) is expressed from human 293 cells (HEK293). It contains AA Gln 35 - Gly 428 (Accession P13688).This protein carrie 1 * 200 µG
Catalog # ACRBCE1-H82E5200UG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,Biotinylated Human CEACAM-1 CD66a Protein, Avitag™,His Tag,Biotinylated Human CEACAM-1, Avitag,His Tag (CE1-H82E 5) is expressed from human 293 cells (HEK293). It contains AA Gln 35 - Gly 428 (Accession P13688).This protein carrie 1 * 200 µG
Catalog # ACRBCE1-H82E5200UG
Supplier:  ACROBIOSYSTEMS