Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
Cynomolgus CD40, His Tag (CD0-C52H6) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Arg 193 (Accession G7PG38) . 1 * 100 µG
ACRBCD0-C52H6100UG
:
ACROBIOSYSTEMS
Cynomolgus CD40, His Tag (CD0-C52H6) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Arg 193 (Accession G7PG38) . 1 * 100 µG
ACRBCD0-C52H6100UG
:
ACROBIOSYSTEMS