ContactUsLinkComponent
Recombinat Protein,Human PTPRD Protein, His Tag,Human PTPRD, His Tag (PTD-H52H9) is expressed from human 293 cells (HEK293). It cont ains AA Glu 21 - Glu 1265 (Accession P23468-1).This protein carries a polyhistidine tag at the C-terminus. The prote 1 * 1 mg
Katalog # ACRBPTD-H52H9-1MG
Leverantör:  ACROBIOSYSTEMS
Recombinat Protein,Human PTPRD Protein, His Tag,Human PTPRD, His Tag (PTD-H52H9) is expressed from human 293 cells (HEK293). It cont ains AA Glu 21 - Glu 1265 (Accession P23468-1).This protein carries a polyhistidine tag at the C-terminus. The prote 1 * 1 mg
Katalog # ACRBPTD-H52H9-1MG
Leverantör:  ACROBIOSYSTEMS