ContactUsLinkComponent
Recombinat Protein,Human GAPDH G3PDH Protein, His Tag,Human GAPDH, His Tag (GAH-H5145) is expressed from E.coli cells. It contains AA Gly 2 - Glu 335 (Accession AAH01601).This protein carries a polyhistidine tag at the N-terminus. The protein has a 1 * 1 mg
Katalog # ACRBGAH-H5145-1MG
Leverantör:  ACROBIOSYSTEMS
Recombinat Protein,Human GAPDH G3PDH Protein, His Tag,Human GAPDH, His Tag (GAH-H5145) is expressed from E.coli cells. It contains AA Gly 2 - Glu 335 (Accession AAH01601).This protein carries a polyhistidine tag at the N-terminus. The protein has a 1 * 1 mg
Katalog # ACRBGAH-H5145-1MG
Leverantör:  ACROBIOSYSTEMS