ContactUsLinkComponent
Biotinylated Human Mesothelin, Fc,Avitag (MSN-H82F6) is expressed from human 293 cells (HEK293). It contains AA Glu 296 - Gly 580 ( Accession AAH09272.1). 1 * 200 µG
Katalog # ACRBMSN-H82F6200UG
Leverantör:  ACROBIOSYSTEMS
Biotinylated Human Mesothelin, Fc,Avitag (MSN-H82F6) is expressed from human 293 cells (HEK293). It contains AA Glu 296 - Gly 580 ( Accession AAH09272.1). 1 * 200 µG
Katalog # ACRBMSN-H82F6200UG
Leverantör:  ACROBIOSYSTEMS