ContactUsLinkComponent
Mouse OX40, His Tag (OX0-M5228) is expressed from human 293 cells (HEK293). It contains AA Val 20 - Pro 211 (Accession P47741-1). 1 * 100 µG
Katalog # ACRBOX0-M5228100UG
Leverantör:  ACROBIOSYSTEMS
Mouse OX40, His Tag (OX0-M5228) is expressed from human 293 cells (HEK293). It contains AA Val 20 - Pro 211 (Accession P47741-1). 1 * 100 µG
Katalog # ACRBOX0-M5228100UG
Leverantör:  ACROBIOSYSTEMS