ContactUsLinkComponent
Human CD28H, His Tag (CDH-H52H3) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Gly 150 (Accession Q96BF3-1). 1 * 1 mg
Katalog # ACRBCDH-H52H3-1MG
Leverantör:  ACROBIOSYSTEMS
Human CD28H, His Tag (CDH-H52H3) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Gly 150 (Accession Q96BF3-1). 1 * 1 mg
Katalog # ACRBCDH-H52H3-1MG
Leverantör:  ACROBIOSYSTEMS