ContactUsLinkComponent
Human PD-L1, Strep Tag (PD1-H5282) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Arg 238 (Accession NP054862. 1). 1 * 1 mg
Katalog # ACRBPD1-H5282-1MG
Leverantör:  ACROBIOSYSTEMS
Human PD-L1, Strep Tag (PD1-H5282) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Arg 238 (Accession NP054862. 1). 1 * 1 mg
Katalog # ACRBPD1-H5282-1MG
Leverantör:  ACROBIOSYSTEMS