ContactUsLinkComponent
Human FGL1, His Tag (FG1-H52Hy) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Ile 312 (Accession Q08830-1). 1 * 1 mg
  ACRBFG1-H52HY-1MG
Human FGL1, His Tag (FG1-H52Hy) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Ile 312 (Accession Q08830-1). 1 * 1 mg
  ACRBFG1-H52HY-1MG