Mouse EGF R, His Tag (EGR-M5224) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 647 (Accession Q01279-1). 1 * 50 µG
Katalog # ACRBEGR-M5224-50UG
Mouse EGF R, His Tag (EGR-M5224) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 647 (Accession Q01279-1). 1 * 50 µG
Katalog # ACRBEGR-M5224-50UG