ContactUsLinkComponent
[EN]CHILLER UNICHILLER 060TW C3 -20-40C 1 * 1 Items
Katalog # HUBE3026.0106.01
Leverantör:  HUBER
[EN]CHILLER UNICHILLER 060TW C3 -20-40C 1 * 1 Items
Katalog # HUBE3026.0106.01
Leverantör:  HUBER