Orders LinkContactUsLinkComponent
ESC (Easy Surface Checking) swabs
ESC (Easy Surface Checking) swabs
ESC swab Buffered Peptone Water, 100 tubes×10 ml
Katalog # 85603.
Leverantör:   LIOFILCHEM
ESC (Easy Surface Checking) swabs
Katalog # 85603.
Leverantör:  LIOFILCHEM
ESC (Easy Surface Checking) swabs
ESC (Easy Surface Checking) swabs
ESC swab Buffered Peptone Water, 100 tubes×10 ml
Price
1 925,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 100 Items
Product Details & Documents
The ESC (Easy Surface Checking) swab range includes a new generation of sampling devices for microbiological examination of surfaces, especially recommended for food and pharmaceutical industries.
  • Pre-filled swab device
  • Rayon bud
  • Wide range of diluents
  • Leak proof closure
  • Transport and storage at room temperature
Certifications: Swabs conform to international standards (ISO 18593, ISO 17604).
Specifikationer
  • Beskrivning:
    ESC swab Buffered Peptone Water, 100 tubes×10 ml
  • Seq:
    02