Fluorite Objectives for Revolve Microscope
U-Plan fluorite objective, 60×, 0,2 mm
Katalog # ECHLJMAG0609
Leverantör: DISCOVER ECHO
Gå till:
Product Details & Documents
Documents
ECHO Meet Revolve
Product Literature
Specifikationer
- Förstoring:60×
- Beskrivning:U-Plan fluorite objective
- Seq:06
- Arbetsavstånd:0,2 mm