FLED fluorescence intermediate attachments
FLED Fluorescence intermediate attachment (FITC)
- Fluorescence intermediate part
- LED excitation module FITC, consisting of 470 nm LED lamp and FITC filter cube
- Light blocking slider
- Fluorescence intermediate part
- LED excitation module FITC, consisting of 470 nm LED lamp and FITC filter cube
- Light blocking slider
Katalog # MOTI1101011400191
Leverantör: MOTIC
Gå till:
Product Details & Documents
Compatible with Panthera U, KU and CC microscopes.
Specifikationer
- Beskrivning:FLED Fluorescence intermediate attachment (FITC)
- Fluorescence intermediate part
- LED excitation module FITC, consisting of 470 nm LED lamp and FITC filter cube
- Light blocking slider