FLED fluorescence intermediate attachments
FLED Fluorescence intermediate attachment (Auramine O)
- Fluorescence intermediate part
- LED excitation module AO, consisting of 455 nm LED lamp and AO filter cube
- Light blocking slider
Note: Recommended to use with Plan objectives UC PL NCG
- Fluorescence intermediate part
- LED excitation module AO, consisting of 455 nm LED lamp and AO filter cube
- Light blocking slider
Note: Recommended to use with Plan objectives UC PL NCG
Katalog # MOTI1101011400181
Leverantör: MOTIC
Gå till:
Product Details & Documents
Compatible with Panthera U, KU and CC microscopes.
Specifikationer
- Beskrivning:FLED Fluorescence intermediate attachment (Auramine O)
- Fluorescence intermediate part
- LED excitation module AO, consisting of 455 nm LED lamp and AO filter cube
- Light blocking slider
Note: Recommended to use with Plan objectives UC PL NCG