BucketBinders™ Multi Bucket Systems
Double bucket system, 2 PP buckets (dark blue and yellow), 1 set of BucketBinder™ connectors
Catalog # 141-1470
Supplier: MICRONOVA
Jump to:
Product Details & Documents
BucketBinders™ bucket system is a cost effective, ergonomic and simple that replaces the need for bulky carts.
- Easy clip action allows attachment before or after bucket is filled with disinfectant
- Polyketone hooks firmly secure two or three rectangular buckets together
- Stainless steel base plate attaches at caster for support
- Sturdy enough to secure two or three buckets and easy to maneuver with up to 15 litres of liquid in each bucket
- Lightweight; less than 1 kg per pair
- Allows scale up or down as required
- Autoclavable
The system offers easy movement, cleaning, and storage. Works well in both small and large controlled environments and the sterile core.
Will house W-94 flat mop Megawringer™, W-9, W-92 flat wringers or W-9B BoxWringer for string mops. Complements all the Micronova M-Zone FlatMops™, SnapMops™ and SlimLine™ mops.
: B-7 polypropylene buckets with autoclavable castors available in five colours, (blue, green, red, green and grey) are available as replacement buckets.
: BucketBinder™ kits include two or three polypropylene buckets with castors (22 L capacity), and one or two sets of BucketBinder bucket connectors.
Specifications
- Beskrivning:Double bucket system, 2 PP buckets (dark blue and yellow), 1 set of BucketBinder™ connectors
- Seq:01
Accessories
Product Information
- Catalog #BeskrivningAvailabilityPrice
- MICRBB-01BucketBinder, includes set of stainless steel and polyketone connectorsLog in for availability1 * 1 SET - 5 400,00 kr