Ethylbenzene solution, NMR reference standard, Supelco®
Ethylbenzene solution, 10% in chloroform-d (99.8 atom % D), NMR reference standard, NMR tube size 5 mm x 8 in., ultra-thin wall, 1
736198-8IN
: Merck
- Koncentration:10% in chloroform-d (99.8 atom % D)
- Förp.:1
- Standard:NMR reference standard
- Beskrivning:Ethylbenzene solution
- Pk Info:NMR tube size 5 mm x 8 in., ultra-thin wall
- Cat. No.:736198-8IN
- Benämning:Ethylbenzene solution
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



