Ethylbenzene solution, NMR reference standard, Supelco®
Ethylbenzene solution, 0.4% in chloroform-d (99.8 atom %), NMR reference standard, NMR tube size 3 mm x 8 in., 10
Katalog # 612758-10X8IN
Leverantör: Merck
Ethylbenzene solution, NMR reference standard, Supelco®
Katalog # 612758-10X8IN
Leverantör: Merck
CAS-nummer:
Gå till:
Product Details & Documents
Specifikationer
Specifikationer
- Koncentration:0.4% in chloroform-d (99.8 atom %)
- Förp.:10
- Standard:NMR reference standard
- Beskrivning:Ethylbenzene solution
- Pk Info:NMR tube size 3 mm x 8 in.
- Cat. No.:612758-10X8IN
- Benämning:Ethylbenzene solution



