Ethylbenzene solution, NMR reference standard, Supelco®
Ethylbenzene solution, 0.1% in chloroform-d (99.8 atom % D), NMR reference standard, NMR tube size 8 mm x 7 in., 10
Katalog # 612456-10X7IN
Leverantör: Merck
Ethylbenzene solution, NMR reference standard, Supelco®
Katalog # 612456-10X7IN
Leverantör: Merck
CAS-nummer:
Gå till:
Product Details & Documents
Specifikationer
Specifikationer
- Koncentration:0.1% in chloroform-d (99.8 atom % D)
- Förp.:10
- Standard:NMR reference standard
- Beskrivning:Ethylbenzene solution
- Pk Info:NMR tube size 8 mm x 7 in.
- Cat. No.:612456-10X7IN
- Benämning:Ethylbenzene solution
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



