Accessories for circulators
4-way header with ball valves, M24×1,5, The 4-way header with universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit. Note: The manifold must not be mounted on the unit.
Katalog # HUBE343310
Leverantör: HUBER
Gå till:
Produktdetaljer och dokument
Specifikationer
- Beskrivning:4-way header with ball valves, M24×1,5
- Seq:24
- För:The 4-way header with universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit. Note: The manifold must not be mounted on the unit.