Accessories for circulators
3-way header with ball valves, M24×1,5, 3-way valve system with universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit.
Note: The manifold must not be mounted on the unit.
Note: The manifold must not be mounted on the unit.
Katalog # HUBE343308
Leverantör: HUBER
Gå till:
Product Details & Documents
Specifikationer
- Beskrivning:3-way header with ball valves, M24×1,5
- Seq:23
- För:3-way valve system with universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit.
Note: The manifold must not be mounted on the unit.