Accessories for circulators
2-way valve system, M24×1,5, 2-way valve system with universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit.
Note: The manifold must not be mounted on the unit.
Note: The manifold must not be mounted on the unit.
Katalog # HUBE343306
Leverantör: HUBER
Gå till:
Product Details & Documents
Specifikationer
- Beskrivning:2-way valve system, M24×1,5
- Seq:22
- För:2-way valve system with universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit.
Note: The manifold must not be mounted on the unit.