Accessories for circulators
2-way header, M24×1,5, The 2-way header with universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit. For a direct connection to the unit an additional elbow connector is required.
Katalog # HUBE343221
Leverantör: HUBER
Gå till:
Product Details & Documents
Specifikationer
- Beskrivning:2-way header, M24×1,5
- Seq:19
- För:The 2-way header with universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit. For a direct connection to the unit an additional elbow connector is required.