Accessories for circulators
5-way header with ball valves, M16×1, The 5-way header with universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit.
Note: The manifold must not be mounted on the unit.
Note: The manifold must not be mounted on the unit.
Katalog # HUBE343305
Leverantör: HUBER
Gå till:
Produktdetaljer och dokument
Specifikationer
- Beskrivning:5-way header with ball valves, M16×1
- Seq:17
- För:The 5-way header with universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit.
Note: The manifold must not be mounted on the unit.