Accessories for circulators
4-way header with ball valves, M38×1,5, The 4-way header with universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit.
Note: The manifold must not be mounted on the unit.
Note: The manifold must not be mounted on the unit.
Katalog # HUBE343331
Leverantör: HUBER
Gå till:
Product Details & Documents
Specifikationer
- Beskrivning:4-way header with ball valves, M38×1,5
- Seq:15
- För:The 4-way header with universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit.
Note: The manifold must not be mounted on the unit.