Accessories for circulators
3-way header with ball valves, M38×1,5, The 3-way header with universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit.
Note: The manifold must not be mounted on the unit.
Note: The manifold must not be mounted on the unit.
Catalog # HUBE343329
Supplier: HUBER
Jump to:
Product Details & Documents
Specifications
- Beskrivning:3-way header with ball valves, M38×1,5
- Seq:14
- För:The 3-way header with universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit.
Note: The manifold must not be mounted on the unit.