Accessories for circulators
4-way header, M38×1,5, The 4-way header with universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit. For a direct connection to the unit an additional elbow connector is required.
Katalog # HUBE343235
Leverantör: HUBER
Gå till:
Produktdetaljer och dokument
Specifikationer
- Beskrivning:4-way header, M38×1,5
- Seq:12
- För:The 4-way header with universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit. For a direct connection to the unit an additional elbow connector is required.