Orders LinkContactUsLinkComponent
Accessories for circulators
Accessories for circulators
3-way header, M16×1, The 3-way header and the TCS should be connected with a suitable length of insulated hose. Universal mounting device for fixing in position is included. For direct connection to the unit, an additional elbow connector is required.
Catalog # HUBE341870
Supplier:   HUBER
Accessories for circulators
Catalog # HUBE341870
Supplier:  HUBER
Accessories for circulators
3-way header, M16×1, The 3-way header and the TCS should be connected with a suitable length of insulated hose. Universal mounting device for fixing in position is included. For direct connection to the unit, an additional elbow connector is required.
Price
4 750,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications
  • Beskrivning:
    3-way header, M16×1
  • Seq:
    02
  • För:
    The 3-way header and the TCS should be connected with a suitable length of insulated hose. Universal mounting device for fixing in position is included. For direct connection to the unit, an additional elbow connector is required.