Orders LinkContactUsLinkComponent
Accessories for circulators
Accessories for circulators
2-way header, M16×1, The 2-way header and the TCS should be connected with a suitable length of insulated hose. Universal mounting device for fixing in position is included. For direct connection to the unit, an additional elbow connector is required.
Katalog # HUBE337657
Leverantör:   HUBER
Accessories for circulators
Katalog # HUBE337657
Leverantör:  HUBER
Accessories for circulators
2-way header, M16×1, The 2-way header and the TCS should be connected with a suitable length of insulated hose. Universal mounting device for fixing in position is included. For direct connection to the unit, an additional elbow connector is required.
Price
3 500,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivning:
    2-way header, M16×1
  • Seq:
    01
  • För:
    The 2-way header and the TCS should be connected with a suitable length of insulated hose. Universal mounting device for fixing in position is included. For direct connection to the unit, an additional elbow connector is required.