Tablet counting triangles, SteriWare®
Sterile, Vit, 230×230×13 mm
Catalog # SAMP8066A-1S
Supplier: Sampling Systems
Jump to:
Product Details & Documents
The SteriWare® counting triangles are ideal for counting round tablets. They are manufactured and packed in a cleanroom and so can be used straight from the bag.
- Made from FDA conforming PS
- BSE/TSE free
- Suitable for left handed and right handed operators
- With an embossed scale to aid operation
Certifications: Conforms to FDA, EU 10/2011 and EC 1935/2004. Sterility Assurance Level (SAL) of 10⁻⁶.
Packaging: Individually bagged in a heat-sealed laminate bag.
Specifications
- Färg:Vit
- Beskrivning:Sterile
- Seq:01
- B×D×H:230×230×13 mm