General purpose gloves, A100
XXL, Svart, 12 pairs/box
Catalog # PRTWA100K8RXXL
Supplier: Portwest
Jump to:
Product Details & Documents
Latex palm dipped 10 gauge glove which provides good tear and abrasion resistance, for secure handling in both wet and dry conditions.
- Cotton work gloves
- ANSI cut level A1
- ANSI abrasion level 2
- Crinkle latex grip offers excellent grip
- 100% breathable seamless liner
Latex foam coating for wet and dry conditions. Open back for ventilation and ergonomic design to reduce hand fatigue.
Certifications: Standards: AS/NZS 2161 .2, AS/NZS 2161 .3, ANSI/ISEA 105: 2016 CUT Level A1, EN388:2016 +A1:2018 - 3.1.4.3.X, EN 420.
Specifications
- Handskstorlek:XXL
- Förp.:12 pairs/box
- Färg:Svart
- Seq:08
- Cat. No.:PRTWA100K8RXXL
- Ej handspecifika/handspecifika:Handspecifika