Orders LinkContactUsLinkComponent
General purpose gloves, A100
General purpose gloves, A100
XXL, Svart, 12 pairs/box
Catalog # PRTWA100K8RXXL
Supplier:   Portwest
General purpose gloves, A100
Catalog # PRTWA100K8RXXL
Supplier:  Portwest
General purpose gloves, A100
General purpose gloves, A100
XXL, Svart, 12 pairs/box
Price
350,00 kr
Log in for availability
1 * 12 Par
Product Details & Documents
Latex palm dipped 10 gauge glove which provides good tear and abrasion resistance, for secure handling in both wet and dry conditions.
  • Cotton work gloves
  • ANSI cut level A1
  • ANSI abrasion level 2
  • Crinkle latex grip offers excellent grip
  • 100% breathable seamless liner
Latex foam coating for wet and dry conditions. Open back for ventilation and ergonomic design to reduce hand fatigue.
Certifications: Standards: AS/NZS 2161 .2, AS/NZS 2161 .3, ANSI/ISEA 105: 2016 CUT Level A1, EN388:2016 +A1:2018 - 3.1.4.3.X, EN 420.
Specifications
  • Handskstorlek:
    XXL
  • Förp.:
    12 pairs/box
  • Färg:
    Svart
  • Seq:
    08
  • Cat. No.:
    PRTWA100K8RXXL
  • Ej handspecifika/handspecifika:
    Handspecifika