Objectives for REVOLVE 3M microscope
Planera apokromat; 0,08 NA, 2×, 6,2 mm
Katalog # ECHLJMAG0020
Leverantör: DISCOVER ECHO
Gå till:
Product Details & Documents
Documents
ECHO Meet Revolve
Produktlitteratur
Specifikationer
- Förstoring:2×
- Beskrivning:Planera apokromat; 0,08 NA
- Seq:02
- Arbetsavstånd:6,2 mm