Fluorite Objectives for Revolve Microscope
Fluorite ELWD objective, 60×, 1,5 - 2,2 mm
Katalog # 630-3063
Leverantör: DISCOVER ECHO
Gå till:
Product Details & Documents
Specifikationer
- Förstoring:60×
- Beskrivning:Fluorite ELWD objective
- Seq:14
- Arbetsavstånd:1,5 - 2,2 mm