Objectives for REVOLVE 3M microscope
Plan achromat LWD fas PhC; 0,4 NA, 20×, 3,2 mm
Katalog # ECHLJMAG0209
Leverantör: DISCOVER ECHO
Gå till:
Product Details & Documents
Documents
ECHO Meet Revolve
Produktlitteratur
Specifikationer
- Förstoring:20×
- Beskrivning:Plan achromat LWD fas PhC; 0,4 NA
- Seq:15
- Arbetsavstånd:3,2 mm