CapLock™ clips
CapLock™ clip, Röd
Katalog # SIMPT330-75CLR
Leverantör: Simport Scientific
Gå till:
Produktdetaljer och dokument
These CapLock™ clips are made of PE. They provide extra security against inadvertent cap openings on critical samples or during incubation above 80 °C.
- Ideal for shipping samples
- Suitable for 5 ml tubes with snap cap
Specifikationer
- Färg:Röd
- Beskrivning:CapLock™ clip
- Seq:67