CapLock™ clips
CapLock™ clip, Blå
Catalog # SIMPT330-75CLB
Supplier: Simport Scientific
Jump to:
Product Details & Documents
These CapLock™ clips are made of PE. They provide extra security against inadvertent cap openings on critical samples or during incubation above 80 °C.
- Ideal for shipping samples
- Suitable for 5 ml tubes with snap cap
Specifications
- Färg:Blå
- Beskrivning:CapLock™ clip
- Seq:65