VWR® Basic, Overalls, with Hood
Blå, L
Leverantör: VWR Collection
Gå till:
Produktdetaljer och dokument
These disposable overalls, with hood, are made of non woven, spunbonded polypropylene (SPP), a lightweight fabric that provides protection in non hazardous, dirty work environments.
- Hood
- Zip closure (one direction)
- Elasticated wrists
- Free from natural rubber latex
Weight: 40 g/m²
Packaging: 1 piece per bag, 50 bags per case.
Documents
Specifikationer
- Klädstorlek:L
- Färg:Blå
- Seq:09
- Cat. No.:113-9022
- Vikt:40 g/m²
- Outfit applikationer:General Purpose|Laboratory