VWR® Basic, Overalls
Blå, XXL
Leverantör: VWR Collection
Gå till:
Product Details & Documents
These disposable overalls, with collar, are made of non woven, spunbonded polypropylene (SPP), a lightweight fabric that provides protection in non hazardous, dirty work environments.
- Collar
- Zip closure (one direction)
- Elasticated wrists
- Free from natural rubber latex
Weight: 40 g/m²
Förpackning: 1 piece per bag, 50 bags per case.
Dokument
CoC VWR Overalls coc_VWRI113-9002_12020256 coc_VWRI113-9002_28320258 coc_VWRI113-9003_05120239 coc_VWRI113-9003_28320258
More
Food contact certificate F
Ytterligare produktinformation
VWR® Non Regulated Disposables for Production
Produktlitteratur
Specifikationer
- Klädstorlek:XXL
- Färg:Blå
- Seq:07
- Cat. No.:113-9008
- Vikt:40 g/m²
- Outfit applikationer:General Purpose|Laboratory