VWR® Basic, Overalls
Blå, XL
Supplier: VWR Collection
Jump to:
Product Details & Documents
These disposable overalls, with collar, are made of non woven, spunbonded polypropylene (SPP), a lightweight fabric that provides protection in non hazardous, dirty work environments.
- Collar
- Zip closure (one direction)
- Elasticated wrists
- Free from natural rubber latex
Weight: 40 g/m²
Packaging: 1 piece per bag, 50 bags per case.
Documents
CoC VWR Overalls coc_VWRI113-9002_12020256 coc_VWRI113-9002_28320258 coc_VWRI113-9003_05120239 coc_VWRI113-9003_28320258
More
Food contact certificate F
Additional Product Details
VWR® Non Regulated Disposables for Production
Product Literature
Specifications
- Klädstorlek:XL
- Färg:Blå
- Seq:06
- Cat. No.:113-9007
- Vikt:40 g/m²
- Outfit applikationer:General Purpose|Laboratory