Ziprasidone ≥98.0%
1 g
Katalog # TCIAZ0034-1G
Leverantör: TCI
Gå till:
Product Details & Documents
Synonymer: Ziprasidonum, 5-(2-(4-(Benzo[d]isothiazol-3-yl)piperazin-1-yl)ethyl)-6-chloroindolin-2-one
- CAS-nummer:146939-27-7
- Formel:C₂₁H₂₁ClN₄OS
- MDL-nummer:MFCD00866661
- Molecular Weight:412.94 g/mol
- Merck Index:14,10171
Specifikationer
Specifikationstestresultat
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 95.0 %
- Melting point:218.0 to 224.0 deg-C
Specifikationer
- Förp.:1 g
- Cat. No.:TCIAZ0034-1G
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



