Ziprasidone ≥98.0%
100 mg
Katalog # TCIAZ0034-100MG
Leverantör: TCI
Gå till:
Product Details & Documents
Synonymer: Ziprasidonum, 5-(2-(4-(Benzo[d]isothiazol-3-yl)piperazin-1-yl)ethyl)-6-chloroindolin-2-one
- CAS-nummer:146939-27-7
- Formel:C₂₁H₂₁ClN₄OS
- MDL-nummer:MFCD00866661
- Molecular Weight:412.94 g/mol
- Merck Index:14,10171
Specifikationer
Specifikationstestresultat
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 95.0 %
- Melting point:218.0 to 224.0 deg-C
Specifikationer
- Förp.:100 mg



