Orders LinkContactUsLinkComponent
Density hydrometers
Density hydrometers
Hydrometer 21, 1,900 - 1,960 g/cm³, 0,001 g/cm³, 300 mm
Katalog # AMARH800174
Leverantör:   Amarell
Density hydrometers
Katalog # AMARH800174
Leverantör:  Amarell
Density hydrometers
Density hydrometers
Hydrometer 21, 1,900 - 1,960 g/cm³, 0,001 g/cm³, 300 mm
Price
685,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Product Details & Documents
General purpose hydrometers for density areas.
  • Paper scale and shot weighted body
  • Hydrometer accuracy ±1 scale division
  • Thermometer inside the hydrometer body
Used in laboratories and industry.
Specifikationer
  • Beskrivning:
    Hydrometer 21
  • Längd:
    300 mm
  • Seq:
    51
  • Indelning:
    0,001 g/cm³
  • Mätområde:
    1,900 - 1,960 g/cm³