Compact burettes, BLAUBRAND®
50 ml, 0,1 ml, 0,05 ± ml
Katalog # 612-7164
Leverantör: Brand
Gå till:
Produktdetaljer och dokument
Borosilicate 3.3 glass, amber, class AS.
- Removable PTFE stopcock, fast to dismantle and easy to clean
- Simple repair - all individual components replaceable
- Calibrated to deliver (TD, Ex)
Certifications: DIN EN ISO 385, DE-M marking
Ordering information: On demand also available with individual certificate, USP Individual certificate or DAkkS calibration certificate.
Delivery information: Delivered with batch number and enclosed one batch certificate per packing unit of the manufacturer.
Specifikationer
- Kapacitet:50 ml
- Seq:03
- Indelning:0,1 ml
- Tolerans:0,05 ± ml