Order Entry
Sweden
Orders LinkContactUsLinkComponent
H1 oat medium

H1 oat medium

H1 oat medium 10 L, 10 L
Katalog # APOSPMO612B-10L
Leverantör:   Apollo Scientific
H1 oat medium
Katalog # APOSPMO612B-10L
Leverantör:  Apollo Scientific
CAS-nummer:  
H1 oat medium

H1 oat medium 10 L, 10 L

CAS-nummer:  
Price
615,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 10 L
Product Details & Documents

Contains agar and sucrose. Contains the macro- and micronutrients and supplements required for the symbiotic culture of orchids.

  • Plant tissue culture tested
Specifikationer

Specifikationer

  • Förp.:
    10 L
  • Beskrivning:
    H1 oat medium 10 L

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.