Orders LinkContactUsLinkComponent
Anderson's Basal salt mixture
Anderson's Basal salt mixture
Anderson's basal salt mixture 10×1 L, 10
Katalog # APOSPMA267A-10X1L
Leverantör:   Apollo Scientific
Anderson's Basal salt mixture
Katalog # APOSPMA267A-10X1L
Leverantör:  Apollo Scientific
CAS-nummer:  
Anderson's Basal salt mixture
Anderson's basal salt mixture 10×1 L, 10
CAS-nummer:  
Price
260,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 10 Items
Product Details & Documents
Contains macro/micronutrients as described byAnderson, 1978/80 Plant tissue culture testedFull specification given in appendix.
  • MFCD00243139
Specifikationer
Specifikationer
  • Förp.:
    10
  • Beskrivning:
    Anderson's basal salt mixture 10×1 L