Human bone marrow, unprocessed
Human unprocessed bone marrow, Fresh, Pooled, 100 mL
Katalog # BM008F-4
Leverantör: Charles River Laboratories Cell Solutions
Human bone marrow, unprocessed
Katalog # BM008F-4
Leverantör: Charles River Laboratories Cell Solutions
CAS-nummer:
Gå till:
Produktdetaljer och dokument
Bone marrow is a major source of stem cells, which are being used in cancer research, cell studies, transplantation studies, and other research.
- Test negative for Hepatitis B and C, HIV 1/2, HTLV- I/II, HCV/HBV, WNV, Trypanasoma cruzi, and Syphilis
Bone marrow collected from healthy human volunteer donors under IRB approved informed consent, drawn from the posterior iliac crest using a syringe containing anticoagulant.
: Intended for research use only and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.
Specifikationer
Specifikationer
- Cellformat:Fresh
- Förp.:100 mL
- Enskild donator/poolad:Pooled
- Beskrivning:Human unprocessed bone marrow