Orders LinkContactUsLinkComponent
Objectives, Panthera
Objectives, Panthera
CCIS® Plan achromatic objective UC PL 100×/1,25/S-Oil (WD=0,16 mm), 100x/1,25 (i), 0,16 mm
Catalog # MOTI1101001705251
Supplier:   MOTIC
Objectives, Panthera
Catalog # MOTI1101001705251
Supplier:  MOTIC
Objectives, Panthera
Objectives, Panthera
CCIS® Plan achromatic objective UC PL 100×/1,25/S-Oil (WD=0,16 mm), 100x/1,25 (i), 0,16 mm
Price
1 900,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications
  • Förstoring:
    100x/1,25 (i)
  • Beskrivning:
    CCIS® Plan achromatic objective UC PL 100×/1,25/S-Oil (WD=0,16 mm)
  • Seq:
    07
  • Arbetsavstånd:
    0,16 mm