Orders LinkContactUsLinkComponent
Ground protection accessories
Ground protection accessories
Ground mat, 8534×1829 mm
Katalog # 119-0318
Leverantör:   JUSTRITE MANUFACTURING
Ground protection accessories
Katalog # 119-0318
Leverantör:  JUSTRITE MANUFACTURING
Ground protection accessories
Ground protection accessories
Ground mat, 8534×1829 mm
Price
10 225,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Product Details & Documents
These ground protection accessories improve berm longevity without sacrificing functionality.
  • Ground mats create a smooth, uniform deployment area with this heavy-duty woven fabric
  • Track mats position inside the berm for extra protection along tire paths
  • Track runners place this high visibility, yellow vinyl coated fabric inside the berm for moderate protection against tire abrasion
Specifikationer
  • Beskrivning:
    Ground mat
  • L×B:
    8534×1829 mm
  • Seq:
    04