Ground protection accessories
Ground mat, 8534×1829 mm
Katalog # 119-0318
Leverantör: JUSTRITE MANUFACTURING
Gå till:
Product Details & Documents
These ground protection accessories improve berm longevity without sacrificing functionality.
- Ground mats create a smooth, uniform deployment area with this heavy-duty woven fabric
- Track mats position inside the berm for extra protection along tire paths
- Track runners place this high visibility, yellow vinyl coated fabric inside the berm for moderate protection against tire abrasion
Specifikationer
- Beskrivning:Ground mat
- L×B:8534×1829 mm
- Seq:04