Ground protection accessories
Track runner, 8534×914 mm
Catalog # 119-0317
Supplier: JUSTRITE MANUFACTURING
Jump to:
Product Details & Documents
These ground protection accessories improve berm longevity without sacrificing functionality.
- Ground mats create a smooth, uniform deployment area with this heavy-duty woven fabric
- Track mats position inside the berm for extra protection along tire paths
- Track runners place this high visibility, yellow vinyl coated fabric inside the berm for moderate protection against tire abrasion
Specifications
- Beskrivning:Track runner
- L×B:8534×914 mm
- Seq:02