Orders LinkContactUsLinkComponent
Ground protection accessories
Ground protection accessories
Track runner, 8534×914 mm
Catalog # 119-0317
Ground protection accessories
Catalog # 119-0317
Ground protection accessories
Ground protection accessories
Track runner, 8534×914 mm
Price
2 425,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
These ground protection accessories improve berm longevity without sacrificing functionality.
  • Ground mats create a smooth, uniform deployment area with this heavy-duty woven fabric
  • Track mats position inside the berm for extra protection along tire paths
  • Track runners place this high visibility, yellow vinyl coated fabric inside the berm for moderate protection against tire abrasion
Specifications
  • Beskrivning:
    Track runner
  • L×B:
    8534×914 mm
  • Seq:
    02